Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Tetanus Toxoid IgG (Rat) ELISA Kit

Catalog No. 89100686 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-686 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-686 Supplier Abnova™ Supplier No. KA2496
Add to Cart
Add to Cart

Includes

  • Tetanus Toxoid coated 96-well plate1, 96 wells (12 strips of 8 wells)
  • Enzyme Conjugate Reagent, 11mL
  • Reference Standard2 (lyophilized), 1 vials, Store ≤-20°C
  • 20x Wash Solution, 50mL
  • Diluent, 30mL
  • TMB Reagent (One-step), 11mL
  • Stop Solution (1N HCl), 11mL

Use to measure quantitatively Tetanus Toxoid IgG in serum.

The assay uses tetanus toxoid for solid phase (microtiter wells) immobilization and horseradish peroxidase (HRP) conjugated anti-rat IgG antibodies for detection

Specifications

Calibration Range 0.39 to 25U/mL
Conjugate HRP
Cross Reactivity Rat
Description Tetanus Toxoid IgG (Rat) ELISA Kit is for the quantitative measurement of Tetanus Toxoid IgG in serum.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store lyophilized standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type Tetanus Toxoid IgG (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.