Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Tetanus Toxoid IgM (Monkey) ELISA Kit

Catalog No. 89100638 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-638 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-638 Supplier Abnova™ Supplier No. KA2443
Add to Cart
Add to Cart

Includes

  • Tetanus toxoid coated wells, 96 wells (12 strips of 8 wells)
  • Enzyme Conjugate Reagent, 11mL
  • Standard stock (lyophilized), The standard was calibrated with affinity purified anti-tetanus toxoid IgM. 2 vials. Store < -20°C
  • 20x Wash Solution, 50mL
  • Diluent, 13mL
  • TMB Reagent, 11mL
  • Stop Solution (1N HCl), 11mL

Use to measure quantitatively Tetanus Toxoid IgM in serum.

The monkey anti-tetanus toxoid IgG ELISA is a solid phase enzyme-linked immunosorbent assay

Specifications

Calibration Range 62.5 to 1000ng/mL
Conjugate HRP
Cross Reactivity Monkey
Description Tetanus Toxoid IgM (Monkey) ELISA Kit is for the quantitative measurement of Tetanus Toxoid IgM in serum.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store lyophilized standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type Tetanus Toxoid IgM (Monkey) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.