Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TF (Human) ELISA Kit

Catalog No. 89100215 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Cell Culture Samples, CSF, Milk, Saliva, Urine
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-215 Plasma, Serum
89-100-214 Cell Culture Samples, CSF, Milk, Saliva, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-215 Supplier Abnova™ Supplier No. KA0509
Add to Cart
Add to Cart

Includes

  • Human Transferrin Microplate: A 96 well polystyrene microplate coated with a polyclonal antibody against human transferrin
  • Sealing Tapes
  • Human Transferrin Standard: Human transferrin in a buffered protein base, lyophilized, 75μg
  • Biotinylated Human Transferrin: Lyophilized
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

This assay employs a quantitative competitive enzyme immunoassay technique that measures human transferring in less than 2 hours

Specifications

Calibration Range 0.098 to 25ug/mL
Conjugate Biotin
Cross Reactivity Human
Description TF (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of human TF in plasma and serum.
Detection Limit 0.07μg/mL
Detection Method Colorimetric
Includes Antibody coated microwells 96 wells, Human TF Standard (60μg), Biotinylated TF 1 vial, MIX Diluent Concentrate (10x) (30mL), Wash Buffer Concentrate (20x) (30mL), Streptavidin-Peroxidase Conjugate (SP Conjugate) 80 Chromogen Substrate (8mL)Stop Solution: (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 25 μL
Packaging 96-well
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.