Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TF (Mouse) ELISA Kit

Catalog No. 89100666 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-666 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-666 Supplier Abnova™ Supplier No. KA2476
Add to Cart
Add to Cart

Includes

  • Anti-transferrin coated 96-wells, 96 wells (12 strips of 8 wells)
  • HRP Conjugate Reagent, 11mL
  • Reference Standard (Lyophilized), 1 vial
  • 20x Wash Solution, 50mL
  • 10x Diluent, 25mL
  • TMB Reagent (One-Step), 11mL
  • Stop Solution (1N HCl), 11mL

Use to measure quantitatively Tf in plasma and serum.

The Tf (Mouse) ELISA Kit is based on a solid phase enzyme-linked immunosorbent assay (ELISA)

Specifications

Conjugate HRP
Cross Reactivity Mouse
Description Tf (Mouse) ELISA Kit is for the quantitative measurement of Tf in plasma and serum.
Detection Limit 3.125 to 200ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Tf (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.