Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Tf (Rat) ELISA Kit

Catalog No. 89100703 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
89-100-703 HRP
89-028-598 Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-703 Supplier Abnova™ Supplier No. KA2513
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sandwich Enzyme Immunoassay for the quantitative measurement of rat Tf

Specifications

Conjugate HRP
Cross Reactivity Rat
Description Tf (Rat) ELISA Kit is for the quantitative measurement of Tf in plasma and serum.
Detection Limit 3.125 to 100 ng/mL
Detection Method Colorimetric
Includes Component, Anti rat transferrin coated plate, Reference standard 1 vial (Lyophilized), 10x Diluent (25mL), HRP Conjugate Reagent (11mL), 20x Wash Solution (50mL), TMB Reagent (One-step) (11mL), Stop Solution (1N HCl) (11mL)
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Tf (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.