Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TFRC (Human) ELISA Kit

Catalog No. 89022533 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-533 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-533 Supplier Abnova™ Supplier No. KA0039
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Antibody Coated Microtiter Strips ready to use 96 wells
  • Conjugate solution ready to use 13mL
  • Set of standards concentrated 6x0.1mL
  • Quality Control HIGH concentrated 0.05mL
  • Quality Control LOW concentrated 0.05mL
  • Dilution Buffer ready to use 20mL
  • Biotin++Ab Diluent ready to use 13mL
  • Wash Solution Conc. (10x) concentrated 100mL
  • Substrate Solution ready to use 13mL
  • Stop Solution ready to use 13mL
  • Product Data Sheet and Certificate of Analysis - 1 pc

Sandwich Enzyme Immunoassay for the quantitative measurement of Human soluble transferrin receptor in Serum, Plasma and tissue culture medium

Specifications

Calibration Range 0.05 to 2μg/mL
Conjugate HRP
Cross Reactivity Human
Description TFRC (Human) ELISA Kit is a double monoclonal sandwich enzyme immunoassay for the quantitative measurement of human soluble transferrin receptor in serum, plasma and tissue culture medium.
Detection Limit 2ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microtiter strip, 13mL Conjugate solution, 6 x 0.1mL Set of standards, 0.05mL Quality control high, 0.05mL Quality control low, 2 x 13mL Dilution Buffer, 13mL Biotin-Ab Diluent, 100mL wash solution Conc. (10X), 13mL Substrate solution, 13mL stop solution
Quantity 1 Kit
Sample Type Plasma (Citrate, EDTA, Heparin), Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type TFRC (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.