Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TGFB1 (Human) ELISA Kit

Catalog No. 89101122 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Volume:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume
89-101-122 50 μL
89-100-809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-122 Supplier Abnova™ Supplier No. KA3826
Add to Cart
Add to Cart

Includes

  • Human LAP TGF-β1 Microplate: A 96 well polystyrene microplate coated with a murine monoclonal antibody against LAP TGF-β1, 96 wells
  • Sealing Tapes
  • Human LAP TGF-β1 Standard: Human LAP TGF-β1 in a buffered protein base (lyophilized), 2ng
  • Biotinylated Human LAP TGF-β1 Antibody (50x)
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate, 80μL
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

This assay employs a quantitative sandwich enzyme immunoassay technique that measures LAP TGF-β1 in less than 5 hours

Specifications

Conjugate Biotin
Cross Reactivity Human
Description TGFB1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human TGFB1.
Detection Limit 0.03ng/mL
Detection Method Colorimetric
Includes Human LAP TGFβ1 Microplate, Sealing Tapes, Human LAP TGFβ1 Standard (2ng), Biotinylated Human LAP TGFβ1 Antibody (50x) (140μL), MIX Diluent Concentrate (10x) (30mL), Wash Buffer Concentrate (20x) (30mL x 2), Streptavidin-Peroxidase Conjugate (SP Conjugate) (80μL), Chromogen Substrate: 8mL, Stop Solution: (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Milk, Plasma, Serum
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store the kit at 4°C. For long term storage, please store at -20°C.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.