Learn More
Description
THBS1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human THBS1
Quantification
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Human |
| Description | THBS1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human THBS1. |
| Quantity | 1 Kit |
| Content And Storage | Store the kit at -20°C |
| For Use With (Application) | Quantification |
| Product Type | THBS1 (Human) ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.