Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Thromboxane B2 ELISA Kit

Catalog No. 89100175 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-175 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-175 Supplier Abnova™ Supplier No. KA0294
Add to Cart
Add to Cart

Includes

  • Goat anti-Rabbit IgG Microtiter Plate, 96 Wells
  • TXB2 ELISA Conjugate, 6mL
  • TXB2 ELISA Antibody, 6mL
  • Assay Buffer, 30mL
  • Wash Buffer Concentrate, 30mL
  • Thromboxane B2 Standard, 0.5mL
  • pNpp Substrate, 20mL
  • Stop Solution, 5mL
  • Plate Sealer, 1 slice

Use to measure quantitatively Thromboxane B2 in biological fluids. A competitive immunoassay.

The Thromboxane B2 ELISA kit is a competitive immunoassay for the quantitative determination of Thromboxane B2 in biological fluids

Specifications

Calibration Range 13.7 to 10,000pg/mL
Conjugate AP
Description Thromboxane B2 ELISA Kit is a competitive immunoassay for the quantitative determination of Thromboxane B2 in biological fluids.
Detection Limit 10.54pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Biological fluid
Sample Volume 100 μL
Sensitivity 10.54 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.