Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Thyroglobulin Ab ELISA Kit

Catalog No. 89027257 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-257 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-257 Supplier Abnova™ Supplier No. KA1119
Add to Cart
Add to Cart

For the quantitative measurement of IgG class autoantibodies against Thyroglobulin in Human Serum or Plasma

Specifications

Calibration Range 100 to 9000IU/mL
Conjugate HRP
Description Thyroglobuline-Ab ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against thyroglobulin (TG) in human serum or plasma.
Detection Limit 10U/mL
Detection Method Colorimetric
Includes 1.5mL Combined calibrator with IgG class Anti-TG Antibodies, 1.5mL Each Anti-TG controls 2 vials, 20mL Sample Buffer 1 vial, 15mL Enzyme Conjugate solution 1 vial, 15mL TMB Substrate solution 1 vial, 15mL stop solution 1 vial, 96-well plate 1 Plate, 20mL
Quantity 96 Tests
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Thyroglobulin Ab ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.