Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Thyroglobulin (Human) ELISA Kit

Catalog No. 89027259 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-259 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-259 Supplier Abnova™ Supplier No. KA1121
Add to Cart
Add to Cart

Indirect Solid Phase ELISA for the quantitative measurement of Thyroglobulin in Human Serum

Specifications

Calibration Range 3 to 300ng/mL
Conjugate HRP
Description Thyroglobulin (hTG) ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of thyroglobulin in human serum.
Detection Limit 1ng/mL
Detection Method Colorimetric
Includes 1.5mL Each Thyroglobulin calibrator 6 Vials, 1.5mL Each Thyroglobulin controls 2 Vials, 3mL Thyroglobulin recovery control 1 Vial, 15mL Sample Buffer 1 Vial, 15mL Enzyme Conjugate solution 1 Vial, 15mL TMB Substrate solution 1 Vial, 96-well plate 1 Plate, 15mL stop solution 1 Vial, 20mL Wash solution 1 Vial
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Thyroglobulin (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.