Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Thyroglobulin IgG ELISA Kit

Catalog No. 89028715 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-715 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-715 Supplier Abnova™ Supplier No. KA1899

Anti-TG ELISA Kit is an enzyme immunoassay for the quantitative measurement of anti-TG autoantibodies in human serum.

Quantification

Specifications

Conjugate HRP
Cross Reactivity Human
Detection Limit 10IU/mL
Detection Method Colorimetric
Includes Dilution plate 1 plate, 1 x 50mL Wash Buffer concentrate, 1 x 12mL Substrate, 1 x 12mL stop solution, 1 x 1mL calibrator 0, 1 x 1mL calibrator 1, 1 x 1mL calibrator 2, 1 x 1mL calibrator 3, 1 x 1mL calibrator 4, 1 x 1mL calibrator 5, TG-coated microtiter
Quantity 96 Assays
Sample Type Serum
Sample Volume 10 μL
Content And Storage 4°C
For Use With (Application) For the quantitative measurement of Anti-TG autoantibodies in Human Serum
Product Type Thyroglobulin IgG ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.