Learn More
Specifications
Specifications
| Conjugate | HRP |
| Description | Tick borne encephalitis virus IgG ELISA Kit is a solid phase enzyme immunoassay for the semiquantitative measurement of IgG class autoantibodies against Tick borne encephalitis virus. |
| Detection Method | Colorimetric |
| Includes | 96-well ELISA break away strip coated with specific Antigen 96-well, 1.3mL negative control Serum, 1.3mL positive control Serum, 0.2mL Antibodies-Px Conjugate, 125mL Wash Buffer concentrate, 125mL Dilution Buffer, 15mL Chromogenic Substrate, 30mL stop solution |
| Quantity | 1 Kit |
| Sample Type | Serum |
| Content And Storage | Store at 4°C on dry atmosphere away from light. |
| For Use With (Application) | Quantification |
| Product Type | Tick borne encephalitis Virus IgG ELISA kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.