Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TIMP1 (Human) ELISA Kit

Catalog No. 89025038 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
89-025-038 HRP
89-100-814 Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-025-038 Supplier Abnova™ Supplier No. KA1199

Sandwich Enzyme Immunoassay for the quantitative measurement of Human TIMP-1

Specifications

Calibration Range 0.156 to 10ng/mL
Conjugate HRP
Cross Reactivity Human
Detection Limit 0.078ng/mL
Detection Method Colorimetric
Includes Assay plate, 20mL standard, 20mL sample diluent, 10mL assay diluent A, 10mL assay diluent B, 120μL detection reagent A, 120μL detection reagent B, 30mL Wash Buffer (25X), 10mL Substrate, 10mL stop solution, plate sealer
Quantity 96 Tests
Regulatory Status For research use only (RUO)
Sample Type Biological Fluid, CCS, Plasma, Serum
Sample Volume 100 μL
Content And Storage 4°C
For Use With (Application) For the quantitative measurement of Human TIMP1
Product Type TIMP1 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.