Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TIMP3 (Human) ELISA Kit

Catalog No. 89022856 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-856 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-856 Supplier Abnova™ Supplier No. KA0420
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-human TIMP-3 antibody, 96 (8x12) wells
  • Lyophilized recombinant human TIMP-3 standard, 10ng x 2
  • Biotinylated anti-human TIMP-3 antibody, dilution 1:100, 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL
  • Adhesive cover, 4 slides

Use to measure quantitatively human TIMP3. Sandwich enzyme immunoassay.

TIMP3 (Human) ELISA Kit was based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Antigen TIMP3
Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description TIMP3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human TIMP3.
Detection Limit 2pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, lyophilized recombinant Human TIMP3 standard 10ng x 2 vials, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human TIMP3 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin-biotin
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Saliva
Sample Volume 100 μL
Sensitivity <2 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type ELISA Kit
Specificity Natural and recombinant human TIMP-3
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.