Learn More
Abnova™ Tissue Acetyl-Histone H3 ChIP Kit
Catalog No. 89027030 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-027-030 1 Kit
1 options
Catalog No. 89-027-030 Supplier Abnova™ Supplier No. KA0671
Add to Cart
Add to Cart
Tissue Acetyl-Histone H3 ChIP Kit is for investigating the interactions of histone H3 acetylation and DNA in variety of mammalian tissues.
  • The fastest procedure available, which can be finished within 5 hours
  • Strip microwell format makes the assay flexible: manual or high throughput
  • Columns for DNA purification are included: save time and reduce labor
  • Compatible with all DNA amplification-based approaches
  • Simple, reliable, and consistent assay conditions
  • Upon receipt store the following components at 4°C: Protease Inhibitor Cocktail , Normal Mouse IgG, Anti-Acetyl-Histone H3, Proteinase K, and 8-Well Assay Strips; store all other components at room temperature
  • The kit is stable for up to 6 months from the shipment date, when stored properly
  • This kit is for research use only and is not intended for diagnostic or therapeutic application
Specifications
Description Tissue Acetyl-Histone H3 ChIP Kit is for investigating the interactions of histone H3 acetylation and DNA in variety of mammalian tissues.
Content And Storage Store the following components at 4°C: Protease inhibitor cocktail, normal mouse IgG, acetyl-histone H3 antibody, proteinase K, 8 well strips. Store other components at room temperature.
Quantity 1 Kit
For Use With (Application) Chromatin Immunoprecipitation
Type ChIP Kit