Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Tissue Methylated DNA Immunoprecipitation Kit

Catalog No. 89027050 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-050 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-050 Supplier Abnova™ Supplier No. KA0693
Add to Cart
Add to Cart

Tissue Methylated DNA Immunoprecipitation Kit is used to enrich methylated DNA from various mammalian tissues.

  • Directly immunoprecipitate the methylated fractions of DNA from cell lysates
  • Highly efficient enrichment of methylated DNA: > 95%
  • The fastest procedure available, which can be finished within 4 hours
  • Strip microplate format makes the assay flexible: manual or high throughput
  • Columns for DNA purification are included: save time and reduce labor
  • Compatible with all DNA amplification-based approaches
  • Simple, reliable, and consistent assay conditions
  • Upon receipt store the following components at 4°C: Protease inhibitor cocktail, normal mouse IgG, anti-5-methylcytosine, proteinase K, control primers, 8-well assay strips; store all other components at room temperature
  • The kit is stable for up to 6 months from the shipment date, when stored properly
  • This kit has been configured for research use only and is not for diagnostic and clinical use

Specifications

Description Tissue Methylated DNA Immunoprecipitation Kit is used to enrich methylated DNA from various mammalian tissues.
Content And Storage Store the following components at 4°C: Normal mouse IgG, anti-5-methylcytosine, proteinase K, 8 well strips. Store other components at room temperature.
Quantity 1 Kit
For Use With (Application) Methylated DNA Immunoprecipitation
Type Immunoprecipitation/Co-Immunoprecipitation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.