Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Tissue Total RNA Purification Kit

Catalog No. 89028633 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-633 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-633 Supplier Abnova™ Supplier No. U0066
Add to Cart
Add to Cart

Tissue Total RNA Purification Kit is designed specifically for purifying total RNA from a variety of animal and paraffin-embedded tissue.

  • Tissue samples can be efficiently homogenized in a microcentrifuge tube using the provided micropestle
  • Detergents and chaotropic salt are used to lyse cells and inactivate RNase with an optional in-column DNase treatment
  • The procedure does not require phenol extraction or alcohol precipitation and can be completed within 15 minutes
  • The purified RNA is ready for use in RT-PCR, Northern Blotting, Primer Extension and cDNA Library Construction
  • Stores dry at room temperature (15-25°C) for at least 9 months without showing any reduction in performance
  • For research use only

Specifications

Content And Storage Store at room temperature.
Description Tissue Total RNA Purification Kit is designed specifically for purifying total RNA from a variety of animal and paraffin-embedded tissue.
For Use With (Application) Purification
Includes 60mL RB Buffer, 50mL W1 Buffer, 25mL Wash Buffer (add 100mL Ethanol), 15mL RNase-free Water, filter column, RB column, 2mL collection tube, micropestle
Product Type Tissue Total RNA Purification Kit
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.