Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TNF (Human) ELISA Kit

Catalog No. 89100815 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-815 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-815 Supplier Abnova™ Supplier No. KA3113
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human TNF

Specifications

Calibration Range 15.6 to 1000pg/mL (Body Fluid, Tissue Lysate, Cell Culture Supernatant), 7.8 to 500pg/mL (Serum)
Conjugate Biotin
Cross Reactivity Human
Description TNF (Human) ELISA Kit is intended for the quantitative measurement of human TNF.
Detection Limit 1pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Human TNF-alpha antibody, Lyophilized Human TNFα standards: 10ng x 2, Sample / Standard diluent buffer: 30mL, Biotin conjugated anti-Human TNFα antibody (Concentrated Dilution: 1:100): 130μL, Antibody diluent buffer: 12mL, Avidin-Biotin-Peroxidase Complex (ABC) (ConcentratedDilution: 1:100): 130μL, ABC diluent buffer: 12mL, TMB substrate: 10mL, Stop solution: 10mL, Wash buffer (25X): 30mL
Quantity 96 Assays
Sample Type Body Fluid, Cell Culture Supernate, Serum, Tissue Lysate
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type TNF (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.