Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TNF (Rat) ELISA Kit

Catalog No. 89100817 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-817 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-817 Supplier Abnova™ Supplier No. KA3115
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Rat TNFα antibody, 96 (8 x 12) wells
  • Lyophilized TNFα standards, 10ng x 2
  • Sample/Standard diluent buffer, 30mL
  • Biotin conjugated anti-Rat TNFα antibody (Concentrated), Dilution: 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer (25X), 30mL

Use to measure quantitatively rat Tnf.

This kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 15.6 to 1000pg/mL (Body Fluid, Tissue Lysate, Cell Culture Supernatant), 7.8 to 501pg/mL (Serum)
Conjugate Biotin
Cross Reactivity Rat
Description Tnf (Rat) ELISA Kit is intended for the quantitative measurement of rat Tnf.
Detection Limit 1pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Rat TNFα antibody 96 (8x12) wells, Lyophilized anti-Rat TNFα standards (10ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Rat TNFα antibody (Concentrated), Dilution: 1:100 (130μL) Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
Sample Volume 100 μL
Sensitivity <1 pg/mL
Packaging 96-well
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type Tnf (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.