Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Tnfrsf11a (Mouse) ELISA Kit

Catalog No. 89028739 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-739 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-739 Supplier Abnova™ Supplier No. KA2007
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Tnfrsf11a

Specifications

Conjugate Biotin
Cross Reactivity Mouse
Description Tnfrsf11a (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Tnfrsf11a.
Detection Limit <2pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, lyophilized recombinant Mouse Tnfrsf11a standard 10ng x 2 vials, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Mouse Tnfrsf11a Antibody, 12mL Antibody Diluent Buffer, 130μL Avidi
Quantity 1 Kit
Sample Type Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type TNFRSF11A (Mouse) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.