Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TNFRSF11B (Human) ELISA Kit

Catalog No. 89100003 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
89-100-003 HRP
89-100-803 Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-003 Supplier Abnova™ Supplier No. KA0030
Add to Cart
Add to Cart

Includes

  • Antibody Coated Microtiter Strips, 96 wells
  • Biotin Labelled Antibody, 13mL
  • Streptavidin-HRP Conjugate, 13mL
  • Master Standard, 1 vial
  • Quality Control HIGH, 1 vial
  • Quality Control LOW, 1 vial
  • Dilution Buffer, 13mL
  • Wash Solution Conc. (10x), 100mL
  • Substrate Solution, 13mL
  • Stop Solution, 13mL

  • The kit measures osteoprotegerin in serum and plasma (EDTA, citrate, heparin)
  • Quality Controls are human serum based
  • Standard is recombinant protein based
  • Components of the kit are provided ready to use, concentrated or lyophilized

Specifications

Calibration Range 1.5 to 60pmol/L
Conjugate HRP
Cross Reactivity Human, Monkey
Description TNFRSF11B (Human) ELISA Kit is a biotin labelled antibody based sandwich enzyme immunoassay for the quantitative measurement of human osteoprotegerin in serum, plasma, synovial fluid and tissue culture medium.
Detection Limit 0.03pmol/L
Detection Method Colorimetric
Includes Antibody Coated Microtiter Strips 96 wells, Biotin Labelled Antibody (13mL), Streptavidin-HRP Conjugate (13mL), Master Standards 1 vial, Quality Control HIGH 1 vial, Quality Control LOW 1 vial, Dilution Buffer (13mL), Wash Solution Conc. (10x) (100mL) Substrate Solution (13mL), Stop Solution (13mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma (Citrate, EDTA, Heparin), Serum
Sample Volume 100 μL
Sensitivity 0.03 pmol/L
Total Assay Time <3.5 hr.
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
Specificity 120pmol/L
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.