Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TNNI2 (Rat) ELISA Kit

Catalog No. 89100701 Shop All Abnova Corporation Products
Change view
Click to view available options
No. of Assays:
96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Assays
89-100-701 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-701 Supplier Abnova™ Supplier No. KA2511
Add to Cart
Add to Cart

Includes

  • Anti SkM-TnI-coated wells, 96 wells
  • SkM-TnI Stock (Lyophilized mouse SkM-TnI), Reconstitute with 0.10mL H2O
  • Standard Diluent, 50mL
  • Sampls Diluent, 25mL
  • Anti SkM-TnI HRP Conjugate, 11mL
  • 20x Wash Solution, 50mL
  • TMB Reagent, 11mL
  • Stop Solution (1N HCl), 11mL

Use to measure quantitatively Tnni2 in plasma and serum.

This ELISA kit uses a detection antibody that is specific for the fast twitch isoform of troponin-I, thereby allowing specific evaluation of skeletal muscle injury

Specifications

Conjugate HRP
Cross Reactivity Rat
Description Tnni2 (Rat) ELISA Kit is for the quantitative measurement of Tnni2 in plasma and serum.
Detection Limit 0.78 to 50ng/mL
Detection Method Colorimetric
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store lyophilized standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type Tnni2 (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.