Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TNNI3 (Monkey), HRP, ELISA Kit

Catalog No. 89100641 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-641 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-641 Supplier Abnova™ Supplier No. KA2446
Add to Cart
Add to Cart

Includes

  • anti cTnI-coated wells 96 wells (provided as 12 detachable strips of 8)
  • cTnI Stock Lyophilized monkey cTnI cTnI Diluent 25mL
  • cTnI HRP Conjugate 11mL
  • 20x Wash Solution 50mL
  • TMB Reagent 11mL
  • Stop Solution (1N HCl) 11mL

For quantitative measurement of IgM in plasma and serum

Specifications

Conjugate HRP
Cross Reactivity Monkey
Description TNNI3 (Monkey) ELISA Kit is for the quantitative measurement of TNNI3 in plasma and serum.
Detection Limit 0.039 to 2.5 ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma and serum
Sample Volume 200 μL
Content And Storage Store lyophilized standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type TNNI3 (Monkey) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.