Learn More
Description
TNNI3 (Pig) ELISA Kit is an enzyme immunoassay for the quantitative determination of TNNI3 in pig plasma
Quantification
Specifications
Specifications
| Conjugate | HRP |
| Description | TNNI3 (Pig) ELISA Kit is for the quantitative measurement of TNNI3 in plasma (EDTA). |
| Content And Storage | Store lyophilized standard at -20°C. Store all other components at 4°C. |
| For Use With (Application) | Quantification |
| Product Type | TNNI3 (Pig) ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.