Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TNNI3 (Rabbit) ELISA Kit

Catalog No. 89100675 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Serum
Serum,Plasma
Sample Volume:
100 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Sample Volume
89-100-675 Serum 100 μL
89-100-677 Serum,Plasma 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-675 Supplier Abnova™ Supplier No. KA2485
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative measurement of TNNI3 in serum

Specifications

Conjugate HRP
Description TNNI3 (Rabbit) ELISA Kit is for the quantitative measurement of TNNI3 in serum.
Detection Method Colorimetric
Includes Anti cTnI-coated wells (96 wells), cTnI Stock: Lyophilized bovine cTnI (Reconstitute with 0.40 ml H2O), cTnI Diluent (12mL), cTnI HRP Conjugate (11mL), 20x Wash Solution (50mL), TMB Reagent (HRP substrate solution) (11mL), Stop Solution (1N HCl) (11mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Content And Storage Store lyophilized standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type TNNI3 (Rabbit) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.