Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Tnni3 (Rat), HRP, ELISA Kit

Catalog No. 89100690 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Plasma and serum
Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-690 Serum
89-100-691 Plasma and serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-690 Supplier Abnova™ Supplier No. KA2500
Add to Cart
Add to Cart

Includes

  • anti cTnI-coated wells 96 wells
  • cTnI Stock: Lyophilized bovine cTnI Reconstitute with 0.40mL H2O
  • cTnI Diluent 12mL
  • cTnI HRP Conjugate 11mL
  • 20x Wash Solution 50mL
  • TMB Reagent: HRP substrate solution 11mL
  • Stop Solution (1N HCl) 11mL

For quantitative measurement of TNNI3 in plasma and serum

Specifications

Calibration Range 0.156 to 10ng/mL
Conjugate HRP
Cross Reactivity Rat
Description Tnni3 (Rat) ELISA Kit is for the quantitative measurement of Tnni3 in serum.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Content And Storage Store lyophilized standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type Tnni3 (Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.