Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TNNT2 (Human) ELISA Kit

Catalog No. 89100997 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-997 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-997 Supplier Abnova™ Supplier No. KA3319

Includes

  • Human anti-Troponin T antibody microtiter plate: A plate using break-apart strips coated with a mouse monoclonal antibody specific to human Troponin T
  • Washing Buffer, 20x
  • Standard Protein: Native human Troponin T (Lyophilized)
  • Standard/Sample Dilution Buffer
  • Secondary Antibody: Biotinylated anti human Troponin T antibody (Lyophilized)
  • Streptavidin HRP
  • Secondary antibody/Streptavidin HRP Dilution Buffer
  • Substrate (Stabilized chromogen): Tetramethylbenzidine (TMB) solution
  • Stop Solution: 1N solution of sulfuric acid (H2SO4)
  • Plate sealers: Adhesive sheet

Use to measure quantitatively human TNNT2. Sandwich enzyme immunoassay.

The design of this assay is based on a sandwich Enzyme-Linked Immunosorbent Assay (ELISA)

Specifications

Calibration Range 62.5 to 4000pg/mL
Conjugate Biotin
Cross Reactivity Human
Detection Limit 209pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Buffered Solution, Cell Lysate, Culture Supernatant, Plasma, Serum
Sample Volume 100 μL (Prediluted)
Sensitivity 209 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
Product Type ELISA Kit
Specificity Human cardiac Troponin I, cardiac Troponin C and Muscle Troponin I
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.