Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Total Histone H3 Acetylation Detection Fast Kit (Fluorometric)

Catalog No. 89028233 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-233 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-233 Supplier Abnova™ Supplier No. KA1539
Add to Cart
Add to Cart

Total Histone H3 Acetylation Detection Fast Kit (Fluorometric) is used for measuring total acetylation of histone H3.

  • Quick and efficient procedure, which can be finished within 2.5 hours
  • Innovative fluorometric assay without the need for radioactivity, electrophoresis, and chromatography
  • Captures histone H3 acetylated at any lysine site with the detection limit as low as 0.4 ng/well and detection range from 5 ng-2 μg/well of histone extracts
  • The control is conveniently included for the quantification of the amount of acetylated histone H3
  • Strip microplate format makes the assay flexible: manual or high throughput
  • Simple, reliable, and consistent assay conditions
  • Upon receipt store F3, F4, and the Standard Control at -20°C; store all other components at 4°C away from light
  • The kit is stable for up to 6 months from the shipment date, when stored properly
  • For research use only

Specifications

Description Total Histone H3 Acetylation Detection Fast Kit (Fluorometric) is used for measuring total acetylation of histone H3.
Product Type Total Histone H3 Acetylation Detection Fast Kit (Fluorometric)
Sample Type Histone Extract
Specificity Histone extracts from mammalian cells/ tissues
For Use With (Application) Functional Study
Quantity 1 Kit
Sample Volume 50 μL
Storage Requirements Store F3, F4, and the Standard Control at -20°C away from light; Store all other components at 4°C away from light. The kit is stable for up to 6 months from the shipment date, when stored properly. Check if buffers, F1 and F2, contain salt precipitates before using. If so, warm (at room temperature or 37°C) and shake the buffers until the salts are redissolved.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.