Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Total Histone H4 Acetylation Detection Fast Kit (Fluorometric)

Catalog No. 89028235 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-235 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-235 Supplier Abnova™ Supplier No. KA1541
Add to Cart
Add to Cart

Total Histone H4 Acetylation Detection Fast Kit (Fluorometric) is used for measuring total acetylation of histone H4.

  • Quick and efficient procedure, which can be finished within 2.5 hours
  • Innovative fluorometric assay without the need for radioactivity, electrophoresis, or chromatography
  • Captures histone H4 acetylated at any lysine site with the detection limit as low as 0.4 ng/well and detection range from 5 ng-2 μg/well of histone extracts
  • The control is conveniently included for the quantification of the amount of acetylated histone H4
  • Strip microplate format makes the assay flexible: manual or high throughput
  • Simple, reliable, and consistent assay conditions
  • Upon receipt store F3, F4 and Standard Control at -20°C; store all other components at 4°C away from light
  • The kit is stable for up to 6 months from the shipment date, when stored properly
  • For research use only

Specifications

Description Total Histone H4 Acetylation Detection Fast Kit (Fluorometric) is used for measuring total acetylation of histone H4.
Product Type Total Histone H4 Acetylation Detection Fast Kit (Fluorometric)
Specificity Histone extracts from mammalian cells/ tissues
For Use With (Application) Functional Study
Quantity 1 Kit
Storage Requirements Store F4, the detection antibody and the standard at -20°C away from light. Store all other components at 4°C away from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.