Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TP53 Designer Transcription Factor Assay Kit

Catalog No. 89024631 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-631 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-631 Supplier Abnova™ Supplier No. KA1349

missing translation for 'validMainframeMsg'

TP53 Designer Transcription Factor Assay Kit is a method for detecting specific transcription factor DNA-binding activity in nuclear extracts.

Functional Study

Specifications

Immunoassay Kit Format ELISA (Standard)
Product Type TP53 designer transcription facto r assay kit
Sufficient For 100 Reactions
For Use With (Application) Functional study, for detecting specific transcription facto r DNA-binding activity in nuclear extract in nuclear extract
Quantity Each
Storage Requirements -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.