Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TPO IgG ELISA Kit

Catalog No. 89028716 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-716 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-716 Supplier Abnova™ Supplier No. KA1900

Enzyme Immunoassay for the quantitative measurement of Anti-TPO autoantibodies in Human Serum

Specifications

Conjugate HRP
Cross Reactivity Human
Description Anti-TPO ELISA Kit is an enzyme immunoassay for the quantitative measurement of anti-TPO autoantibodies in human serum.
Detection Limit 2IU/mL
Detection Method Colorimetric
Includes Dilution plate 1 plate, 1 x 50mL Wash Buffer concentrate, 1 x 12mL Substrate, 1 x 12mL stop solution, 1 x 1mL calibrator 0, 1 x 1mL calibrator 1, 1 x 1mL calibrator 2, 1 x 1mL calibrator 3, 1 x 1mL calibrator 4, 1 x 1mL calibrator 5, TPO-coated microtiter strip 1 x 96 well, 1 x 12mL Conjugate, 1 x 1mL Control Serum, 1 x 12mL sample diluent 1, 1 x 12mL sample diluent 2
Quantity 1 Kit
Sample Type Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type TPO IgG ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.