Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Treponema pallidum Total ELISA Kit

Catalog No. 89028049 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-049 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-049 Supplier Abnova™ Supplier No. KA0970

For the detection of IgG, IgM and IgA antibody to Treponema pallidum in Human Serum or Plasma

Specifications

Conjugate HRP
Description Treponema pallidum Total ELISA Kit is used for the detection of IgG, IgM and IgA antibody to Treponema pallidum in human serum or plasma.
Detection Method Colorimetric
Includes Microwell 12 x 8 x 1, 22mL sample diluent, 1.5mL calibrator, 1.5mL positive control, 1.5mL negative control, 12mL Enzyme Conjugate, 12mL TMB chromogenic Substrate, 12mL stop solution, 25mL Washing concentrate (20X)
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Treponema pallidum Total ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.