Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Trf (Mouse) ELISA Kit

Catalog No. 89101129 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma, Serum
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-129 CCS, Plasma, Serum
89-028-644 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-129 Supplier Abnova™ Supplier No. KA3833
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of TRF in Mouse biological samples

Specifications

Conjugate Biotin
Cross Reactivity Mouse
Description Trf (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Trf.
Detection Method Colorimetric
Includes Mouse Transferrin Microplate: A 96 well polystyrene microplate coated with a polyclonal antibody against mouse transferrin, Sealing Tapes: Each kit contains 3 precut, pressure sensitive sealing tapes that can be cut to fit the format of the individual assay (3 ), Mouse Transferrin Standard: Mouse transferrin in a buffered protein base (lyophilized) (40ng), Biotinylated Mouse Transferrin Antibody (50x): A 50-fold concentrated biotinylated polyclonal antibody against transferrin (140μL), MIX Diluent Concentrate (10x): A 10-fold concentrated buffered protein base (30mL), Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant (30mL x 2 ), Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate (80μL), Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine (8mL), Stop Solution: A 0.5 N hydrochloric acid to stop the chromogen substrate reaction (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum
Sample Volume 50 μL
Content And Storage Store the kit at 4°C. For long term storage, please store at -20°C.
For Use With (Application) Quantification
Product Type TRF (Mouse) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.