Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Tryptophan ELISA Kit

Catalog No. 89028726 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-726 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-726 Supplier Abnova™ Supplier No. KA1916
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enzyme Immunoassay for the quantitative determination of Tryptophan

Specifications

Conjugate HRP
Description Tryptophan ELISA Kit is an enzyme immunoassay for the quantitative determination of tryptophan.
Detection Limit 1.2μg/mL
Detection Method Colorimetric
Includes 96-well reaction plate, adhesive foil 1 Pack, 1 x 20mL Wash Buffer concentrate, 1 x 12mL Enzyme Conjugate, 1 x 12mL Substrate, 1 x 12mL stop solution, 1 x 4mL standard A, 1 x 4mL standard B, 1 x 4mL standard C, 1 x 4mL standard D, 1 x 4mL standard E,, 1 x 4mL standard F, 1 x 6mL Tryptophan AntiSerum, 1 x 20mL Assay Buffer, Equalizing reagent 1 vials, Tryptophan microtiter strip 1 x 96-well, 1 x 4mL D-reagent, 1 x 4mL Control 1, 1 x 4mL Control 2, 1 x 4mL Precipitating reagent, 1 x 20mL Q-Buffer, 1 x 20mL PBS
Quantity 1 Kit
Sample Type Plasma, Serum, Urine
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Tryptophan ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.