Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TSH (Cat) ELISA Kit

Catalog No. 89100573 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-573 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-573 Supplier Abnova™ Supplier No. KA2314
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Antibody-coated microtiter wells, 96 wells/plate
  • Lyophilized Standards (0, 1, 2.5, 5, 10, 25ng/mL), reconstitute in 1.0mL standard diluent, 1 set
  • Enzyme Conjugate Reagent, 12mL
  • TMB color reagent (ready for use), 12mL
  • 20x Wash buffer, 20mL
  • Stop solution (2N HCl), 6mL
  • Standard/Sample Diluent, 20mL

Use to measure quantitatively thyroid stimulating hormone (TSH) in cat serum, plasma. Immunoassay.

The TSH (Cat) ELISA Kit is based on the principle of a solid phase enzyme-linked immunosorbent assay

Specifications

Calibration Range 0 to 25ng/mL
Conjugate HRP
Cross Reactivity Cat
Description TSH (Cat) ELISA Kit is an immunoassay for the quantitative determination of thyroid stimulating hormone (TSH) in cat serum, plasma.
Detection Limit 1ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Serum, Plasma
Sample Volume 100 μL
Sensitivity 1 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.