Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TUBB3 IgG (Human) ELISA Kit

Catalog No. 89028312 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-312 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-312 Supplier Abnova™ Supplier No. KA1472

Indirect Solid Phase ELISA for the semi-quantitative measurement of IgG class autoantibodies against TUBB3

Specifications

Conjugate HRP
Detection Method Colorimetric
Includes 96-well ELISA break away strip coated with specific Antigen 96-well, 1.3mL negative control Serum, 1.3mL positive control Serum, 0.2mL Antibodies-Px Conjugate, 125mL Wash Buffer concentrate, 125mL Dilution Buffer, 15mL Chromogenic Substrate, 30mL stop solution
Quantity 96 Assays
Sample Type Cerebrospinal Fluid, Serum
Sample Volume 100 μL
Content And Storage 4°C
For Use With (Application) For the semi-quantitative measurement of IgG class autoantibodies against TUBB3
Product Type TUBB3 IgG (Human) ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.