Learn More
Specifications
Specifications
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Includes | 96-well ELISA break away strip coated with specific Antigen 96-well, 1.3mL negative control Serum, 1.3mL positive control Serum,1.3mL calibrator, 0.2mL Antibodies-Px Conjugate, 125mL Wash Buffer concentrate, 125mL Dilution Buffer, 15mL Chromogenic Substrate, 30mL stop solution |
| Quantity | 1 Kit |
| Sample Type | Cerebrospinal Fluid, Serum |
| Sample Volume | 100 μL |
| Content And Storage | 4°C |
| For Use With (Application) | For the semi-quantitative measurement of IgM class autoantibodies against TUBB3 |
| Product Type | TUBB3 IgM (Human) ELISA kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.