Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Varicella zoster virus IgG ELISA Kit

Catalog No. 89028296 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CSF, Serum
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-296 CSF, Serum
89-100-524 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-296 Supplier Abnova™ Supplier No. KA1456
Add to Cart
Add to Cart

Includes

  • 8-well break-away strips coated with specific antigen, 96 (8 x 12) wells
  • Standard A 60 mIU (international (mili-) units), r.t.u., Ready to use, 1.3mL
  • Standard B 200 mIU, Ready to use, 1.3mL
  • Standard C 500 mIU, Ready to use, 1.3mL
  • Standard D 1500 mIU, Ready to use, 1.3mL
  • Standard E 10000 mIU, Ready to use, 1.3mL
  • Anti-human IgG antibodies labelled with horseradish peroxidase, Ready to use, (Px-conjugate), 15mL
  • Wash buffer, 10x concentarted, 125mL
  • Dilution buffer, Ready to use, 125mL
  • Chromogenic substrate (TMB substrate), Ready to use, 15mL
  • Stop solution, Ready to use, 15mL
  • Sealable pouch for unused strips, 1 piece

Indirect Solid Phase ELISA for the quantitative measurement of IgG class autoantibodies against Varicella zoster Virus

Specifications

Calibration Range 60 to 10000mIU
Conjugate HRP
Description Varicella zoster virus IgG ELISA Kit is a solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Varicella zoster virus.
Detection Method Colorimetric
Includes 96-well ELISA break away strip coated with specific Antigen 96-well, 1.3mL/ each vial standard A to F, 0.2mL Antibodies-Px Conjugate, 125mL Wash Buffer concentrate, 125mL Dilution Buffer, 15mL Chromogenic Substrate, 30mL stop solution
Quantity 1 Kit
Sample Type CSF, Serum
Sample Volume 100 μL
Content And Storage Store at 4°C on dry atmosphere away from light.
For Use With (Application) Quantification
Product Type Varicella zoster virus IgG ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.