Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Varicella zoster virus IgG ELISA Kit

Catalog No. 89100524 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CSF, Serum
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-524 Plasma, Serum
89-028-296 CSF, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-524 Supplier Abnova™ Supplier No. KA2262
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 8-well break-away strips coated with specific antigen, 96 (8 x 12) wells
  • Standard A 60 mIU (international (mili-) units), r.t.u., Ready to use, 1.3mL
  • Standard B 200 mIU, Ready to use, 1.3mL
  • Standard C 500 mIU, Ready to use, 1.3mL
  • Standard D 1500 mIU, Ready to use, 1.3mL
  • Standard E 10000 mIU, Ready to use, 1.3mL
  • Anti-human IgG antibodies labelled with horseradish peroxidase, Ready to use, (Px-conjugate), 15mL
  • Wash buffer, 10x concentarted, 125mL
  • Dilution buffer, Ready to use, 125mL
  • Chromogenic substrate (TMB substrate), Ready to use, 15mL
  • Stop solution, Ready to use, 15mL
  • Sealable pouch for unused strips, 1 piece

Indirect Solid Phase ELISA for the quantitative measurement of IgG class autoantibodies against Varicella zoster Virus

  • The Varicella zoster virus IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) designed for the quantitative measurement of IgG class antibodies against Varicella zoster virus in human serum and cerebrospinal fluid.
  • Assay Type: Quantitative
  • Detection Method: Colorimetric
  • Label: HRP-conjugate
  • Suitable Sample Types: Serum, Cerebrospinal Fluid
  • Sample Volume: 100μL
  • Regulation Status: For research use only

Specifications

Conjugate HRP
Cross Reactivity Human
Description Varicella zoster virus IgG ELISA Kit is intended for the detection of IgG antibody to VZV in human serum or plasma.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Varicella zoster virus IgG ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.