Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ VCAM1 (Mouse) ELISA Kit

Catalog No. 89100818 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma (EDTA, Heparin), Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-818 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-100-271 CCS, Plasma (EDTA, Heparin), Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-818 Supplier Abnova™ Supplier No. KA3116
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Mouse VCAM-1 antibody, 96 (8 x 12) wells
  • Lyophilized VCAM-1 standards, 10ng x 2
  • Sample/Standard diluent buffer, 30mL
  • Biotin conjugated anti-Mouse VCAM-1 antibody (Concentrated), Dilution: 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer (25X), 30mL

This kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 156 to 10000ng/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Vcam1 (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Vcam1.
Detection Limit 5pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Mouse VCAM-1 antibody 96 (8x12) wells, Lyophilized VCAM-1 standards (10ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Mouse VCAM-1 antibody (Concentrated), Dilution: 1:100 (130μL)Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Sensitivity <5 pg/mL
Packaging 96-well
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type Vcam1 (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.