Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ VWF (Human) ELISA Kit

Catalog No. 89022939 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-939 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-939 Supplier Abnova™ Supplier No. KA0512
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • VWF Microplate: A 96-well polystyrene microplate (12 strips of 8 wells) coated with a murine monoclonal antibody against VWF - 1
  • Sealing Tapes: pressure-sensitive sealing tapes, which can be cut to fit the format of the individual assay - 3
  • VWF Standard: Human VWF in a buffered protein base ( lyophilized) 160mU
  • Biotinylated VWF Antibody (50x): A 50-fold concentrated biotinylated polyclonal antibody against VWF 140μL
  • MIX Diluent Concentrate (10x): A 10-fold concentrated buffered protein base 30mL
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant 30mL, 2 bottles
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate 80μL
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine 8mL
  • Stop Solution: A 0.5 N hydroxychloric acid to stop the chromogen substrate reaction 12mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human VWF

Specifications

Calibration Range 2.5 to 80mU/mL
Conjugate Biotin
Cross Reactivity Human
Description VWF (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human VWF.
Detection Limit 2.5mU/mL
Detection Method Colorimetric
Includes Human VWF microplate 96 (8x12) well, sealing tapes 3 Slices, Human VWF standard 80 mU, 100μL Biotinylated Human VWF Antibody (80X), 30mL Mix Diluent concentrate (10X), 30mL x 2 Wash Buffer 20X, 80μL Streptavidin-peroxid
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type VWF (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.