Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 28MSM5P1
Encompass_Preferred
Product Type: Antibody. Application: Agglutination, IF, IHC. Conjugate/Tag: Unconjugated. Clone: HE-193. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: KG1 cells or human colorectal carcinoma tissues. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Mixture of erythrocytes of blood group A, glycoprotein fraction isolated from the saliva of secretors with blood group A. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: Multiple.Chromosome: 9q34.2. Gene Symbol: ABO. Gene ID: 28. Ensemble: ENSG00000175164. Uniprot: NP_065202.
Catalog No. 50-188-4752