Learn More
Aviva Systems Biology Corp Abo Antibody - middle region (ARP49519_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49519_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Abo. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: NAGATApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-285-433