Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ ABO/Rh Blood Typing Tray
SCI_ED

Catalog No. S37734 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S37734 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
DOT: Non regulated
Catalog No. S37734 Supplier Innovating Science™ Supplier No. IS3155
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOT: Non regulated

Contain depression wells to perform ABO and Rh blood-typing.

  • Made of styrene
  • Washable and reusable
  • Quantity: 100/Pk.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) With depression wells to perform ABO and Rh blood-typing
Includes ABO/Rh Blood typing tray package of 100.
Quantity 1
Dimensions (L x W x H) 7.25 x 4.5 x 3.25 in.
Class Size 2 Lab Groups
Product Type ABO/Rh Blood Typing Tray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.