Learn More
Description
- Fully automated system for routine multi-chemistry and electrolyte test results
- Uses small sample size (0.1cc of whole blood, serum or plasma) and gives results in approximately 12 minutes
- Self calibrates with every test run and needs no manual data entry
- Full-color touch screen with intuitive user interface
- iQC™ system performs continuous quality control checks on every run, eliminating daily control testing requirements
- Delivers up to 24 individual tests including CLIA-waived lipids, liver enzymes and glucose monitoring
- Advanced technology uses centrifugal and capillary forces to mix diluent, reagents and sample in a self-contained, disposable disc that reduces procedural errors
- Every self-contained reagent disc is barcoded for automatic self-calibration
- Multi- analyte panel results include alpha-numeric patient information, analyte concentrations, reference ranges, sample integrity indices and QC performance
- Easily operated system operates from line or 15V battery power
- Results are printed on a self-adhesive hard copy or via electronic transfer, allowing immediate access to critical information at point-of-care
- Large memory capacity stores up to 5000 patient records
- Externally accessible bi-directional USB data transfer technology facilitates EMR connectivity
Specifications
Specifications
| Type | Abaxis Piccolo Xpress Chemistry Analyzer |
| Sample Type | Whole Blood |
| For Use With (Application) | For routine multi-chemistry and electrolyte test results |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.