Specifications
| Type | Metlyte 8 Panel |
| For Use With (Application) | For italics (begin)in vitroitalics (end) quantitative determination of: BUN, CK, Cl-, CRE, GLU, K+, Na+, tCO2 |
| For Use With (Equipment) | Piccolo Blood Chemistry Analyser or Piccolo xpress™ Chemistry Analyser |
| Quantity | 10/Pk. |