Learn More
Aviva Systems Biology Corp ABP1 Antibody - C-terminal region (ARP41908_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41908_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ABP1. Alternate names: ABP, AOC1, DAO, DAO1, KAO, ABP1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58042