Learn More
Waters Corp Nut, ABPR Gland Packing
Supplier:  Waters Corp 700008881
Encompass_Preferred
This product is a gland packing nut designed for use in various Supercritical Fluid Chromatography (SFC) environments.
  • Designed for ABPR 200 gland packing.
  • Compatible with Prep SFC 150 Mgm environment.
  • Compatible with Prep SFC 150 AP environment.
  • Compatible with SFC 350 environment.
  • Compatible with ABPR-200 environment.
Catalog No. NC3198413