Learn More
Aviva Systems Biology Corp Abra Antibody - C-terminal region (ARP52675_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52675_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Abra. It was validated on Western Blot. . Alternate names: C130068O12Rik, STARSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-753